siRNA Details
Gene Symbol: Fam53b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107556.1 | NP_001101026.1 | 5 | 1669 | ||
XM_008759886.1 | XP_008758108.1 | 4 | 3549 | |||
XM_008759887.1 | XP_008758109.1 | 4 | 5477 | |||
XM_008759888.1 | XP_008758110.1 | 4 | 5438 |
