siRNA Details
| Gene Symbol: Fank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008347.1 | NP_001008348.1 | 10, 11 | 1047 | ||
| XM_006230439.2 | XP_006230501.1 | 10, 11 | 1112 | |||
| GenBank mRNA | BC082095.1 | - | 10, 11 | 1047 | 7134234 | |

Fank1
Genomic Map