siRNA Details
| Gene Symbol: Mrpl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107567.1 | NP_001101037.1 | 4 | 513 | ||
| XM_008760120.1 | XP_008758342.1 | 4 | 415 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl21| Gene Symbol: Mrpl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107567.1 | NP_001101037.1 | 4 | 513 | ||
| XM_008760120.1 | XP_008758342.1 | 4 | 415 | |||