siRNA Details
| Gene Symbol: Frmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008348.1 | NP_001008349.1 | 2, 3 | 247 | ||
| GenBank mRNA | BC085896.1 | - | 2, 3 | 247 | 7193163 | |
| FQ211651.1 | - | 2, 3 | 286 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frmd8| Gene Symbol: Frmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008348.1 | NP_001008349.1 | 2, 3 | 247 | ||
| GenBank mRNA | BC085896.1 | - | 2, 3 | 247 | 7193163 | |
| FQ211651.1 | - | 2, 3 | 286 | |||