siRNA Details
| Gene Symbol: Atl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044241.1 | NP_001037706.1 | 2 | 478 | ||
| XM_006230812.2 | XP_006230874.1 | 2 | 669 | |||
| GenBank mRNA | DQ450893.1 | - | 2 | 478 | ||
| FQ221502.1 | - | 2 | 292 | |||
| FQ226220.1 | - | 2 | 295 | |||
| FQ230464.1 | - | 2 | 546 | |||
| FQ230893.1 | - | 2 | 546 | |||

Atl3
Genomic Map