siRNA Details
| Gene Symbol: Vps13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100975.2 | NP_001094445.2 | 58, 59 | 8236 | ||
| GenBank mRNA | AY325251.1 | - | 15, 16 | 3382 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps13a| Gene Symbol: Vps13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100975.2 | NP_001094445.2 | 58, 59 | 8236 | ||
| GenBank mRNA | AY325251.1 | - | 15, 16 | 3382 | ||