siRNA Details
| Gene Symbol: Gldc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107583.1 | NP_001101053.1 | 2 | 504 | ||
| XM_006231210.2 | XP_006231272.1 | 2 | 530 | |||
| XM_006231211.2 | XP_006231273.1 | 2 | 531 | |||
| Gene Symbol: LOC100911814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749086.3 | XP_003749134.2 | 2 | 531 | ||

Gldc
Genomic Map