siRNA Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014030.1 | NP_001014052.1 | 5 | 861 | ||
| XM_006231506.2 | XP_006231568.1 | 5 | 1412 | |||
| XM_006231507.2 | XP_006231569.1 | 5 | 1096 | |||
| XM_008760432.1 | XP_008758654.1 | 7 | 1553 | |||
| XM_008760433.1 | XP_008758655.1 | 4 | 1013 | |||
| XM_008760434.1 | XP_008758656.1 | 4 | 1330 | |||
| GenBank mRNA | BC083852.1 | - | 5 | 861 | 7123646 | |

Tmem180
Genomic Map