siRNA Details
| Gene Symbol: Cpeb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760379.1 | XP_008758601.1 | 9 | 3223 | ||
| XM_008760380.1 | XP_008758602.1 | 9 | 3154 | |||
| XM_008760381.1 | XP_008758603.1 | 9 | 2715 | |||
| XM_008760382.1 | XP_008758604.1 | 8 | 1934 | |||
| XM_008760383.1 | XP_008758605.1 | 9 | 2069 | |||
| XM_008760384.1 | XP_008758606.1 | 9 | 3223 | |||

Cpeb3
Genomic Map