siRNA Details
| Gene Symbol: Trim39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213562.1 | NP_998727.1 | 2, 3 | 567 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim39| Gene Symbol: Trim39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213562.1 | NP_998727.1 | 2, 3 | 567 | ||