siRNA Details
| Gene Symbol: Ubash3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107620.1 | NP_001101090.1 | 13 | 2004 | ||
| XM_006256215.2 | XP_006256277.1 | 14 | 2128 | |||
| XM_008772818.1 | XP_008771040.1 | 14 | 1744 | |||
| XM_008772819.1 | XP_008771041.1 | 14 | 1739 | |||

Ubash3a
Genomic Map