siRNA Details
| Gene Symbol: Msl3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014032.1 | NP_001014054.1 | 1 | 1275 | ||
| XM_006256504.2 | XP_006256566.1 | 3 | 2857 | |||
| GenBank mRNA | BC079056.1 | - | 1 | 1275 | 7115468 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msl3l2| Gene Symbol: Msl3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014032.1 | NP_001014054.1 | 1 | 1275 | ||
| XM_006256504.2 | XP_006256566.1 | 3 | 2857 | |||
| GenBank mRNA | BC079056.1 | - | 1 | 1275 | 7115468 | |