siRNA Details
| Gene Symbol: Sec63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107637.1 | NP_001101107.1 | 2 | 399 | ||
| XM_006256570.2 | XP_006256632.1 | 2 | 279 | |||
| XM_006256571.2 | XP_006256633.1 | 2 | 280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec63| Gene Symbol: Sec63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107637.1 | NP_001101107.1 | 2 | 399 | ||
| XM_006256570.2 | XP_006256632.1 | 2 | 279 | |||
| XM_006256571.2 | XP_006256633.1 | 2 | 280 | |||