siRNA Details
| Gene Symbol: Prdm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107639.2 | NP_001101109.2 | 4 | 759 | ||
| XM_008772991.1 | XP_008771213.1 | 5 | 724 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prdm1| Gene Symbol: Prdm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107639.2 | NP_001101109.2 | 4 | 759 | ||
| XM_008772991.1 | XP_008771213.1 | 5 | 724 | |||