siRNA Details
| Gene Symbol: Qrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014034.1 | NP_001014056.1 | 11 | 1513 | ||
| XM_006256588.2 | XP_006256650.1 | 11 | 1552 | |||
| GenBank mRNA | BC089213.1 | - | 11 | 1513 | 7302358 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Qrsl1| Gene Symbol: Qrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014034.1 | NP_001014056.1 | 11 | 1513 | ||
| XM_006256588.2 | XP_006256650.1 | 11 | 1552 | |||
| GenBank mRNA | BC089213.1 | - | 11 | 1513 | 7302358 | |