siRNA Details
| Gene Symbol: Ddx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077647.1 | NP_001071115.1 | 13 | 984 | ||
| GenBank mRNA | GQ243745.1 | - | 12 | 764 | ||
| JQ013737.1 | - | 12 | 772 | |||
| S75275.1 | - | 13 | 984 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx4| Gene Symbol: Ddx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077647.1 | NP_001071115.1 | 13 | 984 | ||
| GenBank mRNA | GQ243745.1 | - | 12 | 764 | ||
| JQ013737.1 | - | 12 | 772 | |||
| S75275.1 | - | 13 | 984 | |||