siRNA Details
| Gene Symbol: Osmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005384.1 | NP_001005384.1 | 17 | 2661 | ||
| XM_006232012.2 | XP_006232074.1 | 17 | 2679 | |||
| GenBank mRNA | AB167522.1 | - | 17 | 2661 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osmr| Gene Symbol: Osmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005384.1 | NP_001005384.1 | 17 | 2661 | ||
| XM_006232012.2 | XP_006232074.1 | 17 | 2679 | |||
| GenBank mRNA | AB167522.1 | - | 17 | 2661 | ||