siRNA Details
Gene Symbol: Mtmr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012077.1 | NP_001012077.1 | 2, 3 | 207 | ||
XM_006232046.2 | XP_006232108.1 | 2, 3 | 298 | |||
GenBank mRNA | BC089876.1 | - | 2, 3 | 207 | 7325487 |
