siRNA Details
Gene Symbol: Dcun1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107668.1 | NP_001101138.1 | 2 | 333 | ||
XM_003749267.3 | XP_003749315.1 | 212 | ||||
XM_006232261.2 | XP_006232323.1 | 3 | 331 | |||
XM_006232262.2 | XP_006232324.1 | 2 | 544 | |||
XM_006232263.2 | XP_006232325.1 | 2 | 259 | |||
XM_006232264.2 | XP_006232326.1 | 2 | 999 | |||
XM_008760925.1 | XP_008759147.1 | 2 | 318 | |||
XM_008760926.1 | XP_008759148.1 | 350 | ||||
XM_008760927.1 | XP_008759149.1 | 2 | 1771 |
Gene Symbol: LOC102553014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760957.1 | XP_008759179.1 | 2 | 615 | ||
XM_008760958.1 | XP_008759180.1 | 2 | 210 | |||
XM_008760959.1 | XP_008759181.1 | 2 | 999 | |||
XM_008760960.1 | XP_008759182.1 | 3 | 350 | |||
XM_008760961.1 | XP_008759183.1 | 3 | 335 | |||
XM_008760962.1 | XP_008759184.1 | 2 | 1773 |
