siRNA Details
| Gene Symbol: Rarres1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014790.1 | NP_001014790.1 | 3, 4 | 542 | ||
| GenBank mRNA | AY936478.1 | - | 3, 4 | 542 | ||
| BC105631.1 | - | 3, 4 | 542 | 7386690 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rarres1| Gene Symbol: Rarres1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014790.1 | NP_001014790.1 | 3, 4 | 542 | ||
| GenBank mRNA | AY936478.1 | - | 3, 4 | 542 | ||
| BC105631.1 | - | 3, 4 | 542 | 7386690 | ||