siRNA Details
| Gene Symbol: Dclre1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025687.1 | NP_001020858.1 | 5 | 1018 | ||
| XM_006233078.2 | XP_006233140.1 | 4 | 1100 | |||
| XM_006233079.1 | XP_006233141.1 | 5 | 1013 | |||
| XM_006233080.2 | XP_006233142.1 | 3 | 919 | |||
| GenBank mRNA | BC098939.1 | - | 5 | 1018 | 7455348 | |

Dclre1b
Genomic Map