siRNA Details
| Gene Symbol: Wdr77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008771.3 | NP_001008771.2 | 3 | 440 | ||
| GenBank mRNA | AY321348.1 | - | 4 | 383 | ||
| BC097367.1 | - | 3 | 391 | 7443204 | ||
| CB581439.1 | - | 452 | ||||
| FQ213175.1 | - | 3 | 535 | |||
| FQ226328.1 | - | 3 | 435 | |||
| Gene Symbol: LOC100911453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749373.3 | XP_003749421.1 | 3 | 546 | ||

Wdr77
Genomic Map