siRNA Details
| Gene Symbol: Sass6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107719.1 | NP_001101189.1 | 4 | 659 | ||
| XM_006233227.1 | XP_006233289.1 | 4 | 659 | |||
| XM_006233228.1 | XP_006233290.1 | 4 | 659 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sass6| Gene Symbol: Sass6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107719.1 | NP_001101189.1 | 4 | 659 | ||
| XM_006233227.1 | XP_006233289.1 | 4 | 659 | |||
| XM_006233228.1 | XP_006233290.1 | 4 | 659 | |||