siRNA Details
Gene Symbol: Arhgap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009405.1 | NP_001009405.1 | 22 | 2835 | ||
XM_006233239.2 | XP_006233301.1 | 21 | 4635 | |||
XM_006233240.2 | XP_006233302.1 | 21 | 2824 | |||
XM_006233241.2 | XP_006233303.1 | 21 | 3074 | |||
GenBank mRNA | BC087167.1 | - | 22 | 2835 | 7135494 |
