siRNA Details
| Gene Symbol: Tram1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107724.1 | NP_001101194.1 | 1 | 419 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tram1l1| Gene Symbol: Tram1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107724.1 | NP_001101194.1 | 1 | 419 | ||