siRNA Details
Gene Symbol: Sgms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014043.1 | NP_001014065.1 | 6 | 1513 | ||
XM_006233311.2 | XP_006233373.1 | 7 | 2449 | |||
XM_008761501.1 | XP_008759723.1 | 6 | 2365 | |||
GenBank mRNA | BC085803.1 | - | 6 | 1513 | 7111151 | |
DQ071571.2 | - | 5 | 1084 |
