siRNA Details
Gene Symbol: Rbm43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037649.1 | NP_001032738.1 | 3 | 193 | ||
XM_006234167.2 | XP_006234229.1 | 3 | 257 | |||
XM_006234168.2 | XP_006234230.1 | 3 | 1055 | |||
XM_008761743.1 | XP_008759965.1 | 3 | 1059 | |||
XM_008761744.1 | XP_008759966.1 | 3 | 261 | |||
GenBank mRNA | BC107648.1 | - | 3 | 193 | 7930358 |
