siRNA Details
Gene Symbol: Tanc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002854.1 | NP_001002854.1 | 12 | 1849 | ||
XM_006234204.1 | XP_006234266.1 | 11 | 1768 | |||
XM_006234205.2 | XP_006234267.1 | 10 | 4004 | |||
XM_006234207.2 | XP_006234269.1 | 7 | 1966 | |||
XM_008761838.1 | XP_008760060.1 | 10 | 2182 | |||
XM_008761839.1 | XP_008760061.1 | 10 | 2468 | |||
XM_008761840.1 | XP_008760062.1 | 8 | 3100 | |||
GenBank mRNA | AB098072.1 | - | 12 | 1849 |
