siRNA Details
| Gene Symbol: Psmd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025689.1 | NP_001020860.1 | 12 | 1196 | ||
| GenBank mRNA | BC097427.1 | - | 12 | 1196 | ||
| BC158645.1 | - | 12 | 1144 | 8363877 | ||
| FQ228714.1 | - | 12 | 1173 | |||
| FQ228827.1 | - | 12 | 1171 | |||

Psmd14
Genomic Map