siRNA Details
| Gene Symbol: Nostrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024260.1 | NP_001019431.1 | 5 | 328 | ||
| GenBank mRNA | AY032855.1 | - | 4 | 250 | ||
| BC088446.1 | - | 5 | 328 | 7126459 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nostrin| Gene Symbol: Nostrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024260.1 | NP_001019431.1 | 5 | 328 | ||
| GenBank mRNA | AY032855.1 | - | 4 | 250 | ||
| BC088446.1 | - | 5 | 328 | 7126459 | ||