siRNA Details
| Gene Symbol: Tspan18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107750.1 | NP_001101220.1 | 5 | 623 | ||
| XM_006234573.2 | XP_006234635.1 | 5 | 554 | |||
| XM_006234574.2 | XP_006234636.1 | 6 | 696 | |||
| XM_006234575.2 | XP_006234637.1 | 4 | 474 | |||
| XM_008762027.1 | XP_008760249.1 | 4 | 374 | |||

Tspan18
Genomic Map