siRNA Details
| Gene Symbol: Dusp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012089.1 | NP_001012089.1 | 1 | 185 | ||
| GenBank mRNA | BC088205.1 | - | 1 | 185 | 7380627 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp2| Gene Symbol: Dusp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012089.1 | NP_001012089.1 | 1 | 185 | ||
| GenBank mRNA | BC088205.1 | - | 1 | 185 | 7380627 | |