siRNA Details
| Gene Symbol: Bpifb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107791.1 | NP_001101261.1 | 3, 4 | 227 | ||
| XM_006235317.1 | XP_006235379.1 | 12, 13 | 1187 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bpifb6| Gene Symbol: Bpifb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107791.1 | NP_001101261.1 | 3, 4 | 227 | ||
| XM_006235317.1 | XP_006235379.1 | 12, 13 | 1187 | |||