siRNA Details
| Gene Symbol: Bpifa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107792.1 | NP_001101262.1 | 4 | 528 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bpifa5| Gene Symbol: Bpifa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107792.1 | NP_001101262.1 | 4 | 528 | ||