siRNA Details
Gene Symbol: Rprd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098727.1 | NP_001092197.1 | 2, 3 | 653 | ||
XM_006235412.1 | XP_006235474.1 | 2, 3 | 659 | |||
GenBank mRNA | BC168699.1 | - | 2, 3 | 658 | 8362231 | |
DY310142.1 | - | 2, 3 | 653 | |||
DY565371.1 | - | 2, 3 | 429 |
