siRNA Details
| Gene Symbol: Noxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100171.1 | NP_001093641.1 | 3 | 339 | ||
| XM_006233600.2 | XP_006233662.1 | 3 | 626 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Noxa1| Gene Symbol: Noxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100171.1 | NP_001093641.1 | 3 | 339 | ||
| XM_006233600.2 | XP_006233662.1 | 3 | 626 | |||