siRNA Details
Gene Symbol: Nelfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107817.2 | NP_001101287.2 | 11 | 1630 | ||
GenBank mRNA | BC160853.1 | - | 10 | 1752 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nelfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107817.2 | NP_001101287.2 | 11 | 1630 | ||
GenBank mRNA | BC160853.1 | - | 10 | 1752 |