siRNA Details
| Gene Symbol: Ntng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107825.1 | NP_001101295.1 | 1 | 366 | ||
| XM_006233800.2 | XP_006233862.1 | 1 | 366 | |||
| XM_006233804.2 | XP_006233866.1 | 1 | 366 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ntng2| Gene Symbol: Ntng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107825.1 | NP_001101295.1 | 1 | 366 | ||
| XM_006233800.2 | XP_006233862.1 | 1 | 366 | |||
| XM_006233804.2 | XP_006233866.1 | 1 | 366 | |||