siRNA Details
| Gene Symbol: Gtpbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100815.1 | NP_001094285.1 | 9, 10 | 979 | ||
| GenBank mRNA | CA508019.1 | - | 1, 2 | 689 | ||
| CV103632.1 | - | 7, 8 | 645 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtpbp10| Gene Symbol: Gtpbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100815.1 | NP_001094285.1 | 9, 10 | 979 | ||
| GenBank mRNA | CA508019.1 | - | 1, 2 | 689 | ||
| CV103632.1 | - | 7, 8 | 645 | |||