siRNA Details
| Gene Symbol: Wdr91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127298.2 | NP_001120770.2 | 11 | 1646 | ||
| XM_008762775.1 | XP_008760997.1 | 11 | 1748 | |||
| XM_008762776.1 | XP_008760998.1 | 11 | 2047 | |||
| GenBank mRNA | BC166785.1 | - | 11 | 1646 | 7935588 | |

Wdr91
Genomic Map