siRNA Details
| Gene Symbol: Ephb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107857.1 | NP_001101327.1 | 14 | 3188 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ephb6| Gene Symbol: Ephb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107857.1 | NP_001101327.1 | 14 | 3188 | ||