siRNA Details
| Gene Symbol: Zfp956 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014056.1 | NP_001014078.1 | 4 | 719 | ||
| XM_006236417.2 | XP_006236479.1 | 5 | 1047 | |||
| XM_006236418.2 | XP_006236480.1 | 6 | 1080 | |||
| GenBank mRNA | BC089951.1 | - | 4 | 719 | 7312211 | |

Zfp956
Genomic Map