siRNA Details
| Gene Symbol: Zfp775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107859.1 | NP_001101329.1 | 3 | 356 | ||
| XM_006236421.2 | XP_006236483.1 | 3 | 4348 | |||
| XM_006236422.2 | XP_006236484.1 | 3 | 422 | |||
| XM_008762889.1 | XP_008761111.1 | 2 | 4534 | |||

Zfp775
Genomic Map