siRNA Details
| Gene Symbol: Igf2bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY325199.1 | - | 4 | 292 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igf2bp3| Gene Symbol: Igf2bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY325199.1 | - | 4 | 292 | ||