siRNA Details
| Gene Symbol: Igf2bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749741.3 | XP_003749789.1 | 11 | 1495 | ||
| XM_006236474.2 | XP_006236536.1 | 11 | 1369 | |||
| XM_006236475.2 | XP_006236537.1 | 9 | 1306 | |||
| XM_008762933.1 | XP_008761155.1 | 11 | 1130 | |||
| GenBank mRNA | AY325199.1 | - | 3 | 85 | ||
| AY724479.1 | - | 11 | 1459 | |||

Igf2bp3
Genomic Map