siRNA Details
Gene Symbol: Emc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008355.1 | NP_001008356.1 | 8 | 854 | ||
GenBank mRNA | BC085846.1 | - | 8 | 854 | 7189020 | |
FQ220446.1 | - | 8 | 739 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Emc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008355.1 | NP_001008356.1 | 8 | 854 | ||
GenBank mRNA | BC085846.1 | - | 8 | 854 | 7189020 | |
FQ220446.1 | - | 8 | 739 |