siRNA Details
| Gene Symbol: March8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107882.1 | NP_001101352.1 | 3 | 414 | ||
| XM_008763243.1 | XP_008761465.1 | 1 | 89 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

March8| Gene Symbol: March8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107882.1 | NP_001101352.1 | 3 | 414 | ||
| XM_008763243.1 | XP_008761465.1 | 1 | 89 | |||