siRNA Details
| Gene Symbol: March8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107882.1 | NP_001101352.1 | 6 | 751 | ||
| XM_008763243.1 | XP_008761465.1 | 5 | 1272 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

March8| Gene Symbol: March8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107882.1 | NP_001101352.1 | 6 | 751 | ||
| XM_008763243.1 | XP_008761465.1 | 5 | 1272 | |||