siRNA Details
| Gene Symbol: Cacna2d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191751.1 | NP_001178680.1 | 5 | 670 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacna2d4| Gene Symbol: Cacna2d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191751.1 | NP_001178680.1 | 5 | 670 | ||