siRNA Details
| Gene Symbol: Usp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014058.1 | NP_001014080.1 | 7 | 916 | ||
| GenBank mRNA | BC079101.1 | - | 7 | 916 | 7124857 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp18| Gene Symbol: Usp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014058.1 | NP_001014080.1 | 7 | 916 | ||
| GenBank mRNA | BC079101.1 | - | 7 | 916 | 7124857 | |